6,7-Dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:157327-51-0
:MFCD11846189
Formula:C₆H₉Cl₂N₃
MDL Number:MFCD08457576
:194.600 g/mol
Storage Temperature:Ambient
Frequently Bought Together