Order Entry
Italy
ContactUsLinkComponent
tert-Butyl morpholin-2-ylmethylcarbamate 97%
Irritant
tert-Butyl morpholin-2-ylmethylcarbamate 97%
Catalog # APOSOR905155-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl morpholin-2-ylmethylcarbamate 97%
Catalog # APOSOR905155-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.28 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:173341-02-1
Formula:C₁₀H₂₀N₂O₃
MDL Number:MFCD06808588