
Methyl-2-aminobenzo[d]thiazole-7-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:209459-11-0
:
Formula:C₉H₈N₂O₂S
MDL Number:MFCD09263762
:208.2370 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together