Order Entry
Italy
ContactUsLinkComponent
3',2,2-Trimethylpropiophenone 95%
3',2,2-Trimethylpropiophenone 95%
Catalog # APOSOR900478-1G
Supplier:  Apollo Scientific
CAS Number:  
3',2,2-Trimethylpropiophenone 95%
Catalog # APOSOR900478-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.26 g/mol
Storage Temperature:Ambient
CAS Number:50390-49-3
Formula:C₁₂H₁₆O
MDL Number:MFCD03841163