2,3-Dibromo-5-picoline
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,3-Dibromo-5-methylpyridine
- CAS Number:29232-39-1
- Formula:C₆H₅Br₂N
- MDL Number:MFCD04112574
- Molecular Weight:250.92 g/mol
- Boiling Pt:270.8 °C
- Melting Pt:52…56 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: 2,3-Dibromo-5-methylpyridine
- CAS Number:29232-39-1
- Formula:C₆H₅Br₂N
- MDL Number:MFCD04112574
- Molecular Weight:250.92 g/mol
- Boiling Pt:270.8 °C
- Melting Pt:52…56 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III



