
4'-Chloro-2'-hydroxyacetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6921-66-0
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00238557
:170.6
Boiling Pt:247 °C
Melting Pt:27…30 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together