
2-Formylphenylboronic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40138-16-7
:
Formula:C₇H₇BO₃
MDL Number:MFCD00151822
:149.94 g/mol
Melting Pt:115…120 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together