Order Entry
Italy
ContactUsLinkComponent
1,1,4,4-Tetraphenyl-1,3-butadiene 98%
Irritant
1,1,4,4-Tetraphenyl-1,3-butadiene 98%
Catalog # APOSOR8656-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,1,4,4-Tetraphenyl-1,3-butadiene 98%
Catalog # APOSOR8656-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :358.48 g/mol
Melting Pt:202…204 °C
EINECS:215-914-7
 :GHS07
Storage Temperature:Ambient
CAS Number:1450-63-1
Formula:(C₆H₅)₂CCHCHC(C₆H₅)₂
MDL Number:MFCD00004766