
2-Bromo-5-(4-morpholino)pyridine
Catalog # APOSOR8597-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:81…83 °C
:243.1004 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:200064-11-5
Formula:C₉H₁₁BrN₂O
MDL Number:MFCD02681930