Order Entry
Italy
ContactUsLinkComponent
2-Methoxy-5-methyl-3-nitropyridine 98%
Irritant
2-Methoxy-5-methyl-3-nitropyridine 98%
Catalog # APOSOR8579-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methoxy-5-methyl-3-nitropyridine 98%
Catalog # APOSOR8579-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:33252-62-9
Formula:C₇H₈N₂O₃
MDL Number:MFCD09258778