2-Bromo-5-nitro-6-picoline
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 6-Bromo-2-methyl-3-nitropyridine, 2-Bromo-6-methyl-5-nitropyridine
- CAS Number:22282-96-8
- Formula:C₆H₅BrN₂O₂
- MDL Number:MFCD03095219
- Molecular Weight:217.02 g/mol
- Melting Pt:69…70 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 6-Bromo-2-methyl-3-nitropyridine, 2-Bromo-6-methyl-5-nitropyridine
- CAS Number:22282-96-8
- Formula:C₆H₅BrN₂O₂
- MDL Number:MFCD03095219
- Molecular Weight:217.02 g/mol
- Melting Pt:69…70 °C
- Storage Temperature:Ambient



