Order Entry
Italy
ContactUsLinkComponent
4-(3-Methyl-4-nitrophenyl)morpholine
4-(3-Methyl-4-nitrophenyl)morpholine
Catalog # APOSOR8376-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(3-Methyl-4-nitrophenyl)morpholine
Catalog # APOSOR8376-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:139…140 °C
Storage Temperature:Ambient
CAS Number:220679-09-4
Formula:C₁₁H₁₄N₂O₃
MDL Number:MFCD08692422