
1-(Boc-amino)cyclopropanecarboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:88950-64-5
:
Formula:C₉H₁₅NO₄
MDL Number:MFCD00083257
:201.22 g/mol
Melting Pt:177…178 °C
EINECS:230-181-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together