Order Entry
Italy
ContactUsLinkComponent
3-(4-Chlorophenyl)propionic acid 98%
3-(4-Chlorophenyl)propionic acid 98%
Catalog # APOSOR7880-25G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Chlorophenyl)propionic acid 98%
Catalog # APOSOR7880-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :184.62 g/mol
Melting Pt:127…131 °C
Storage Temperature:Ambient
CAS Number:2019-34-3
Formula:C₉H₉ClO₂
MDL Number:MFCD00016555