0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
2,4-Dibenzyloxy-5-pyrimidineboronic acid
2,4-Dibenzyloxy-5-pyrimidineboronic acid
2,4-Dibenzyloxy-5-pyrimidineboronic acid
APOSOR7669-1G
:
Apollo Scientific
:
2,4-Dibenzyloxy-5-pyrimidineboronic acid
APOSOR7669-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
336.150 g/mol
:
GHS07
:
Refrigerator
:
70523-24-9
:
C₁₈H₁₇BN₂O₄
:
MFCD01318938