
4'-Methoxybiphenyl-4-carboxaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52988-34-8
:
Formula:C₁₄H₁₂O₂
MDL Number:MFCD01862517
Melting Pt:98…100 °C
:212.248 g/mol
:GHS07
Storage Temperature:Ambiente
Frequently Bought Together