
2-Cyanobenzeneboronic acid-1,3-propanediol ester
Catalog # APOSOR7188-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:42…45 °C
:187.00 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:172732-52-4
Formula:C₁₀H₁₀BNO₂
MDL Number:MFCD04039012