Order Entry
Italy
ContactUsLinkComponent
1,2,3,4-Tetrahydropyrazino[1,2-a]indolemethanesulphonate 98%
Irritant
1,2,3,4-Tetrahydropyrazino[1,2-a]indolemethanesulphonate 98%
Catalog # APOSOR70091-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,2,3,4-Tetrahydropyrazino[1,2-a]indolemethanesulphonate 98%
Catalog # APOSOR70091-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.34 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:41838-39-5
Formula:C₁₂H₁₆N₂O₃S
MDL Number:MFCD17676108