
4-Iodo-1-methyl-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:39806-90-1
:
Formula:C₄H₅IN₂
MDL Number:MFCD05663860
:208.0000 g/mol
Melting Pt:59…64 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together