


Cycloheptanone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:502-42-1
:
Formula:C₇H₁₂O
MDL Number:MFCD00004159
:112.17 g/mol
Boiling Pt:179…181 °C
Melting Pt:–21 °C
Density:0.956 g/cm³ (25 °C)
Flash Pt:55 °C
UN:1224
ADR:3,III
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together