

N-Methylethylenediamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:109-81-9
:
Formula:C₃H₁₀N₂
MDL Number:MFCD00008165
:74.13 g/mol
Boiling Pt:114…117 °C
Density:0.85 g/cm³ (25 °C)
Flash Pt:41 °C
Storage Temperature:Ambient
EINECS:203-707-4
UN:2734
ADR:8+3,II
:GHS02
:GHS05
Frequently Bought Together