Vai a Intestazione
Vai a Contenuti principale
Vai a Piè di pagina
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-6,7-dihydro-5H-pyrrolo[1,2-a]imidazole
3-Bromo-6,7-dihydro-5H-pyrrolo[1,2-a]imidazole
3-Bromo-6,7-dihydro-5H-pyrrolo[1,2-a]imidazole
3-Bromo-6,7-dihydro-5H-pyrrolo[1,2-a]imidazole
APOSOR6420-250MG
:
Apollo Scientific
:
3-Bromo-6,7-dihydro-5H-pyrrolo[1,2-a]imidazole
APOSOR6420-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
187.05 g/mol
:
48 °C
:
GHS07
:
Ambient
:
914637-88-0
:
C₆H₇BrN₂
:
MFCD08235255