Order Entry
Italy
ContactUsLinkComponent
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-carbaldehyde 95%
Irritant
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-carbaldehyde 95%
Catalog # APOSOR6188-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-carbaldehyde 95%
Catalog # APOSOR6188-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :136.15 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:914637-04-0
Formula:C₇H₈N₂O
MDL Number:MFCD08235198