
1-[(Ammoniooxy)methyl]-2-chlorobenzene chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5555-48-6
:
Formula:C₇H₉Cl₂NO
MDL Number:MFCD01114592
:194.0585 g/mol
Melting Pt:145…148 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together