
8-Bromo-1-octene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2695-48-9
:
Formula:C₈H₁₅Br
MDL Number:MFCD00000275
:191.11 g/mol
Boiling Pt:92 °C/24mmHg
Density:1.139 g/cm³ (25 °C)
Flash Pt:78 °C
EINECS:220-268-4
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together