
6,7-Dihydrocyclopenta[b]pyridin-5-one
Catalog # APOSOR60178-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:133.15 g/mol
Melting Pt:64…66 °C
:GHS07
Storage Temperature:Ambient
CAS Number:28566-14-5
Formula:C₈H₇NO
MDL Number:MFCD07371500