
2-Chloro-3-formylpyridine
:36404-88-3
:
:C₆H₄ClNO
:MFCD01315308
:141.56 g/mol
:46…54 °C
:>110 °C
:GHS07
:Ambient
Frequently Bought Together
Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™
: Honeywell Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
: PanReac AppliChem
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
8 components: 100mg/l each of Al Cr Cu Fe Mn Ba Zn Zr in HNO3 5% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
N,N-Dimethylacetamide 99%, pure
: Thermo Fisher Scientific
Dichloromethane stabilised for residue analysis, for pesticide residue analysis
: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific