Order Entry
Italy
ContactUsLinkComponent
4,4-Dimethoxybutanone
Flammable
4,4-Dimethoxybutanone
Catalog # APOSOR59432-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
4,4-Dimethoxybutanone
Catalog # APOSOR59432-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :132.16 g/mol
Boiling Pt:170…172 °C
Density:0.996 g/cm³ (25 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:226-605-1
UN:1224
ADR:3,III
 :GHS02
CAS Number:5436-21-5
Formula:C₆H₁₂O₃
MDL Number:MFCD00008789