
Ethyl-4-hydroxy-8-methoxyquinoline-3-carboxylate
Catalog # APOSOR59342-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.2466 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:27568-04-3
Formula:C₁₃H₁₃NO₄
MDL Number:MFCD01829412