Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 4-hydroxy-6-methoxyquinoline-3-carboxylate
Ethyl-4-hydroxy-6-methoxyquinoline-3-carboxylate
Ethyl-4-hydroxy-6-methoxyquinoline-3-carboxylate
Ethyl-4-hydroxy-6-methoxyquinoline-3-carboxylate
APOSOR59341-1G
:
Apollo Scientific
:
Ethyl-4-hydroxy-6-methoxyquinoline-3-carboxylate
APOSOR59341-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
247.24 g/mol
:
Ambient
:
77156-78-6
:
MFCD00085617
:
C₁₃H₁₃NO₄
:
MFCD00844092