Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(+)-2-Methylpiperazine
(S)-(+)-2-Methylpiperazine 97%
(S)-(+)-2-Methylpiperazine 97%
APOSOR5877-10G
:
Apollo Scientific
:
(S)-(+)-2-Methylpiperazine 97%
APOSOR5877-10G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
10 g
Seq:
3236
:
:
100.16 g/mol
:
89…91 °C
:
65 °C
:
Ambient
:
1325
:
4.1,III
:
GHS02
:
GHS07
:
74879-18-8
:
C₅H₁₂N₂
:
MFCD00171452