2,3-Dichloropropene 97%
Supplier: Apollo Scientific
About this item
Synonyms: 2,3-Dichloro-1-propene
- CAS Number:78-88-6
- Formula:C₃H₄Cl₂
- MDL Number:MFCD00000943
- Molecular Weight:110.97 g/mol
- Boiling Pt:94 °C
- Melting Pt:–83 °C
- Density:1.209 g/cm³ (25 °C)
- Flash Pt:10 °C
- Storage Temperature:Ambient
- UN:2047
- ADR:3,II
3 Options Available Below
Product Details & Documents
Synonyms: 2,3-Dichloro-1-propene
- CAS Number:78-88-6
- Formula:C₃H₄Cl₂
- MDL Number:MFCD00000943
- Molecular Weight:110.97 g/mol
- Boiling Pt:94 °C
- Melting Pt:–83 °C
- Density:1.209 g/cm³ (25 °C)
- Flash Pt:10 °C
- Storage Temperature:Ambient
- UN:2047
- ADR:3,II
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.APOSOR5736-100G - Specifications:
More
Specifications
Cat. No.APOSOR5736-500G - Specifications:
More
Specifications
Cat. No.APOSOR5736-1KG
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



