Tetrahydropyran-2-methanol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:100-72-1
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00006624
:116.16 g/mol
Boiling Pt:186…189 °C
Density:1.027 g/cm³ (25 °C)
Flash Pt:>76 °C
Storage Temperature:Ambient
Frequently Bought Together