Order Entry
Italy
ContactUsLinkComponent
 
DL-2,3-Diaminopropionic acid hydrochloride 99%
Catalog # APOSOR5494-25G
Supplier:  Apollo Scientific
CAS Number:  
DL-2,3-Diaminopropionic acid hydrochloride 99%
Catalog # APOSOR5494-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :140.57 g/mol
Melting Pt:232 °C (dec.)
Storage Temperature:Ambient
CAS Number:54897-59-5
Formula:C₃H₈N₂O₂
MDL Number:MFCD00012884