Order Entry
Italy
ContactUsLinkComponent
4-Acetamidobenzenesulphonyl chloride 95%
Corrosive
4-Acetamidobenzenesulphonyl chloride 95%
Catalog # APOSOR5468-1KG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
4-Acetamidobenzenesulphonyl chloride 95%
Catalog # APOSOR5468-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :233.68 g/mol
Melting Pt:142…145 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
CAS Number:121-60-8
Formula:C₈H₈ClNO₃S
MDL Number:MFCD00007442