

(1S,2S)-(-)-1,2-Diphenylethylenediamine
:29841-69-8
:
:C₁₄H₁₆N₂
:MFCD00082751
:80…85 °C
:212.29 g/mol
:GHS05
:GHS07
:Ambient
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals