
2-(Bromomethyl)-1,3-dioxolane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4360-63-8
:
Formula:C₄H₇BrO₂
MDL Number:MFCD00003214
:167 g/mol
Boiling Pt:80…82 °C/27mmHg
Density:1.613 g/cm³ (25 °C)
Flash Pt:62 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together