Order Entry
Italy
ContactUsLinkComponent
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one
Irritant
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one
Catalog # APOSOR53058-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one
Catalog # APOSOR53058-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :148.55 g/mol
Boiling Pt:93 °C/2mmHg
Density:1.36 g/cm³ (25 °C)
Flash Pt:129 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:80841-78-7
Formula:C₅H₅ClO₃
MDL Number:MFCD07787494