
(Diethoxyphosphoryl)methyl-4-methylbenzenesulphonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:31618-90-3
:
Formula:C₁₂H₁₉O₆PS
MDL Number:MFCD03412078
Boiling Pt:137 °C/0.2mmHg
Melting Pt:29…31 °C
:322.3100 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together