Order Entry
Italy
ContactUsLinkComponent
4-Cyanophenyl isothiocyanate 97%
Corrosive
Irritant
4-Cyanophenyl isothiocyanate 97%
Catalog # APOSOR5303-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
4-Cyanophenyl isothiocyanate 97%
Catalog # APOSOR5303-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.2 g/mol
Melting Pt:119…123 °C
Storage Temperature:Refrigerator
EINECS:220-323-2
UN:2923
ADR:8+6.1,III
 :GHS05
 :GHS07
CAS Number:2719-32-6
Formula:C₈H₄N₂S
MDL Number:MFCD00041085