Order Entry
Italy
ContactUsLinkComponent
1,8-Octane diisothiocyanate 97%
1,8-Octane diisothiocyanate 97%
Catalog # APOSOR52987-1G
Supplier:  Apollo Scientific
CAS Number:  
1,8-Octane diisothiocyanate 97%
Catalog # APOSOR52987-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.38 g/mol
Storage Temperature:Ambient
CAS Number:56312-14-2
Formula:C₁₀H₁₆N₂S₂
MDL Number:MFCD00041140