
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:174810-09-4
:
Formula:C₅₂H₄₄N₂O₂P₂
MDL Number:MFCD02684551
:790.8655 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together