1,1'-Bis(dicyclohexylphosphino)ferrocene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146960-90-9
:
Formula:C₃₄H₅₂FeP₂
MDL Number:MFCD09038820
Melting Pt:138 °C
:570.5057 g/mol
Storage Temperature:Ambient
Frequently Bought Together