Order Entry
Italy
ContactUsLinkComponent
2-(2-Bromoethyl)-1,3-dioxolane
Toxic
2-(2-Bromoethyl)-1,3-dioxolane
Catalog # APOSOR52533-5G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
2-(2-Bromoethyl)-1,3-dioxolane
Catalog # APOSOR52533-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :181.03 g/mol
Boiling Pt:68…70 °C/8mmHg
Density:1.542 g/cm³ (25 °C)
Flash Pt:65 °C
Storage Temperature:Refrigerator
EINECS:242-551-1
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:18742-02-4
Formula:C₅H₉BrO₂
MDL Number:MFCD00003216