Ethyl-4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride
Catalog # APOSOR52510-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.642 g/mol
Storage Temperature:Ambient
CAS Number:180258-46-2
Formula:C₇H₁₂ClN₃O₂
MDL Number:MFCD01318760