
3-(4-Methoxyphenyl)propan-1-ol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5406-18-8
:
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00002952
:166.22 g/mol
Boiling Pt:164…168 °C/18mmHg
Melting Pt:26 °C
Flash Pt:>113 °C
Storage Temperature:Ambient
EINECS:226-463-0
:GHS07
Frequently Bought Together