Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(-)-1,2,4-Butanetriol
(S)-(-)-1,2,4-Butanetriol 95%
(S)-(-)-1,2,4-Butanetriol 95%
(S)-(-)-1,2,4-Butanetriol 95%
APOSOR5242-1G
:
Apollo Scientific
:
(S)-(-)-1,2,4-Butanetriol 95%
APOSOR5242-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
106.121 g/mol
:
150 °C/0.05mmHg
:
1.19 g/cm³ (25 °C)
:
110 °C
:
GHS07
:
Ambient
:
42890-76-6
:
C₄H₁₀O₃
:
MFCD00063213