Order Entry
Italy
ContactUsLinkComponent
2-(4-Bromobutyl)-1,3-dioxolane
Irritant
2-(4-Bromobutyl)-1,3-dioxolane
Catalog # APOSOR52318-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(4-Bromobutyl)-1,3-dioxolane
Catalog # APOSOR52318-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Density:1.38 g/cm³ (25 °C)
 :209.08 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:87227-41-6
Formula:C₇H₁₃BrO₂
MDL Number:MFCD02093454