Order Entry
Italy
ContactUsLinkComponent
(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole 98+%
Irritant
(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole 98+%
Catalog # APOSOR51901-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole 98+%
Catalog # APOSOR51901-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :308.44 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1203507-02-1
Formula:C₁₉H₂₀N₂S
MDL Number:MFCD28411847