
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14347-78-5
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00003213
:132.16 g/mol
Boiling Pt:72…73 °C
Density:1.062 g/cm³ (25 °C)
Flash Pt:79 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together